Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004013.hg.1 |
TC01000255.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004013.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004013.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150100.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150100.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150100.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150100.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150100.hg.1 |
TC0100007191.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000375102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
ENST00000465062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007191.hg.1 |
NM_022819.3 |
Refseq |