Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258379.1 | NP_001245308.1 | 20 - 21 | 3099 | 143 | |
NM_001258380.1 | NP_001245309.1 | 20 - 21 | 2861 | 143 | ||
NM_001321103.1 | NP_001308032.1 | 21 - 22 | 3483 | 143 | ||
NM_001321104.1 | NP_001308033.1 | 21 - 22 | 3444 | 143 | ||
NM_001321105.1 | NP_001308034.1 | 21 - 22 | 3444 | 143 | ||
NM_001321106.1 | NP_001308035.1 | 21 - 22 | 3432 | 143 | ||
NM_001321107.1 | NP_001308036.1 | 20 - 21 | 3099 | 143 | ||
NM_001321108.1 | NP_001308037.1 | 20 - 21 | 3111 | 143 | ||
NM_003615.4 | NP_003606.3 | 21 - 22 | 3233 | 143 | ||
NR_135541.1 | 20 - 21 | 3257 | 143 | |||
NR_135542.1 | 19 - 20 | 2924 | 143 | |||
NR_135543.1 | 19 - 20 | 2924 | 143 | |||
NR_135544.1 | 20 - 21 | 3257 | 143 | |||
XM_005265598.4 | XP_005265655.1 | 20 - 21 | 3365 | 143 | ||
XM_005265600.4 | XP_005265657.1 | 20 - 21 | 3327 | 143 | ||
XM_006713421.2 | XP_006713484.1 | 21 - 22 | 3471 | 143 | ||
XM_011534256.2 | XP_011532558.1 | 21 - 22 | 3744 | 143 | ||
XM_011534258.1 | XP_011532560.1 | 21 - 22 | 3447 | 143 | ||
XM_011534261.2 | XP_011532563.1 | 21 - 22 | 3709 | 143 | ||
XM_011534263.2 | XP_011532565.1 | 20 - 21 | 3036 | 143 | ||
XM_011534265.1 | XP_011532567.1 | 20 - 21 | 3075 | 143 | ||
XM_017007526.1 | XP_016863015.1 | 22 - 23 | 3368 | 143 | ||
XM_017007527.1 | XP_016863016.1 | 20 - 21 | 3060 | 143 | ||
XM_017007528.1 | XP_016863017.1 | 14 - 15 | 1984 | 143 | ||
XM_017007529.1 | XP_016863018.1 | 14 - 15 | 1966 | 143 | ||
GenBank mRNA | AF047033.1 | - | 21 - 22 | 3233 | 143 | |
AF053755.1 | - | 17 - 18 | 2539 | 143 | ||
AF089726.1 | - | 20 - 21 | 2861 | 143 | ||
CR627428.1 | - | 13 - 14 | 1815 | 143 | ||
EU499349.1 | - | 20 - 21 | 3026 | 143 | ||
EU934246.1 | - | 21 - 22 | 3359 | 143 | ||
EU934247.1 | - | 21 - 22 | 3371 | 143 | ||
EU934248.1 | - | 21 - 22 | 3410 | 143 | ||
EU934249.1 | - | 20 - 21 | 3026 | 143 | ||
EU934250.1 | - | 21 - 22 | 3371 | 143 | ||
FJ178574.1 | - | 20 - 21 | 3184 | 143 | ||
FJ178575.1 | - | 19 - 20 | 2851 | 143 | ||
FJ178576.1 | - | 19 - 20 | 2851 | 143 | ||
GU354307.1 | - | 20 - 21 | 3184 | 143 | ||
GU354308.1 | - | 20 - 21 | 3038 | 143 | ||
GU354309.1 | - | 19 - 20 | 2863 | 143 | ||
GU354310.1 | - | 19 - 20 | 2863 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |