Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
TC08001507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
ENST00000297578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
ENST00000424899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
ENST00000543107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019804.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800177571.hg.1 |
TC0800011372.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000297578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000521645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
ENST00000523866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NM_030780.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102337.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011372.hg.1 |
NR_102338.1 |
Refseq |