Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010039.hg.1 |
ENST00000258534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010039.hg.1 |
ENST00000544152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010039.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010039.hg.1 |
TC12000793.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163597.hg.1 |
TC1200008629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000258534.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000544152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000546729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000549365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000551403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
ENST00000614926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008629.hg.1 |
NM_018370.2 |
Refseq |