Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019404.hg.1 |
TC04001393.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019404.hg.1 |
ENST00000295256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019404.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019404.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177595.hg.1 |
TC0400011348.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000295256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000506331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
ENST00000514774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011348.hg.1 |
NM_014485.2 |
Refseq |