Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_001126063_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8127515 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8127516 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8127517 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8127511 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
220513_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11729573_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11729573_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022640.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022640.hg.1 |
ENST00000370385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022640.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022640.hg.1 |
ENST00000423730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022640.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022645.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022645.hg.1 |
ENST00000370385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022645.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022645.hg.1 |
ENST00000423730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022645.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022646.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022646.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022646.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022642.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022642.hg.1 |
ENST00000370385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022642.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022642.hg.1 |
ENST00000423730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022642.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022644.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022644.hg.1 |
ENST00000370385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022644.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022644.hg.1 |
ENST00000423730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022644.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022639.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022639.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022639.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022641.hg.1 |
TC06001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022641.hg.1 |
ENST00000370385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022641.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022641.hg.1 |
ENST00000423730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022641.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000257765.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000433730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
ENST00000610435.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014313.hg.1 |
NM_030568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202139.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202139.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202139.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202139.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202139.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600202141.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202141.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202141.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202141.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202141.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202146.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202148.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600202140.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202140.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202140.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202140.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202140.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600202143.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202143.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202143.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202143.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202143.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014312.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
ENST00000370388.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
ENST00000471312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
NM_001126063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202144.hg.1 |
TC0600014312.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
220513_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
220513_at |
- |
- |
Human Genome U133A 2.0 Array |
220513_at |
- |
- |
Human Genome U133A 2.0 Array |
220513_PM_at |
- |
- |
Human Genome U95 Set |
82332_at |
- |
- |
Human Genome U95 Set |
82332_PM_at |
- |
- |
Human Genome U133 Set |
220513_PM_at |
- |
- |
Human Genome U133 Set |
220513_at |
- |
- |