Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002352.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002352.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002352.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002352.hg.1 |
TC20000196.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007085.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
NM_032609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
TC2000007085.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
ENST00000376075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
ENST00000490030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148353.hg.1 |
NM_032609.2 |
Refseq |