Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000245121.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000356157.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000585469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000586198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000586198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000586198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000586198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000586198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000588433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000591522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000591522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000591522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000591522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
ENST00000592005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007233.hg.1 |
NM_031303.3 |
Refseq |