Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007825.hg.1 |
ENST00000429271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009096.hg.1 |
ENST00000388829.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009096.hg.1 |
ENST00000388829.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007825.hg.1 |
ENST00000429271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181898.hg.1 |
TC0300011059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181903.hg.1 |
TC0300011059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000419349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000419783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000496791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
ENST00000620890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
NM_000581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
NM_201397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181899.hg.1 |
TC0300011059.hg.1 |
Clariom D TC |