Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009633.hg.1 |
TC01000573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009633.hg.1 |
ENST00000401061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009633.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000477188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000477188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157995.hg.1 |
TC0100008124.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000401061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000477188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000477188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
ENST00000616135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008124.hg.1 |
NM_015506.2 |
Refseq |