Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026916.hg.1 |
TC07001734.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026916.hg.1 |
ENST00000356362.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026916.hg.1 |
ENST00000469408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026916.hg.1 |
ENST00000544995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026916.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187654.hg.1 |
TC0700012159.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000356362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000469408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000478208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000481939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000482157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
ENST00000487277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012159.hg.1 |
NM_019042.4 |
Refseq |