Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
TC01000288.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
ENST00000374627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
ENST00000374630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
ENST00000374632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
ENST00000400191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
ENST00000465676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
NM_004442.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004438.hg.1 |
NM_017449.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374632.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000400191.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000465676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_001309193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_004442.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_017449.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000374632.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000400191.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
ENST00000465676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_001309193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_004442.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
NM_017449.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150819.hg.1 |
TC0100007295.hg.1 |
Clariom D TC |