Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
TC09001674.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
ENST00000245588.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021337.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180649.hg.1 |
TC0900011805.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000334270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000461970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
ENST00000612514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_001205296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011805.hg.1 |
NM_007344.3 |
Refseq |