Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018132.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018132.hg.1 |
ENST00000398675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018132.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018132.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018132.hg.1 |
TC12001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008194.hg.1 |
ENST00000446657.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008194.hg.1 |
ENST00000446657.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010448.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
ENST00000280876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
ENST00000398675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
NM_001099650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
NM_173601.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178740.hg.1 |
TC1200010448.hg.1 |
Clariom D TC |