Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000318837.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000369486.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000481589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
ENST00000481589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001007237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015401.hg.1 |
NM_001542.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008186.hg.1 |
ENST00000625078.1 |
Ensembl |