Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008287.hg.1 |
TC09000686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008287.hg.1 |
ENST00000300456.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008287.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
ENST00000372870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008860.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159424.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159424.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159424.hg.1 |
ENST00000300456.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159424.hg.1 |
NM_005094.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159424.hg.1 |
TC0900008860.hg.1 |
Clariom D TC |