Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000216513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000554079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
ENST00000556952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009353.hg.1 |
NM_017420.4 |
Refseq |