Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01020934.hg.1 |
TC01001330.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01020934.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01020934.hg.1 |
ENST00000490146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01020934.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100174841.hg.1 |
TC0100010198.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000337428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000472465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000476550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
ENST00000490146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010198.hg.1 |
NM_144702.2 |
Refseq |