Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025023.hg.1 |
TC01001619.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025023.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025023.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180744.hg.1 |
TC0100011001.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000367460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000481707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000491030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
ENST00000492967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011001.hg.1 |
NM_130782.2 |
Refseq |