Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173595.3 | NP_775866.2 | 26 - 27 | 2980 | 63 | |
XM_011538197.2 | XP_011536499.1 | 25 - 26 | 3082 | 63 | ||
XM_017019183.1 | XP_016874672.1 | 25 - 26 | 3163 | 63 | ||
GenBank mRNA | AB190990.1 | - | 26 - 27 | 2997 | 63 | |
AK125379.1 | - | 26 - 27 | 2997 | 63 | ||
AK126893.1 | - | 19 - 20 | 2139 | 63 | ||
BC042070.1 | - | 6 - 7 | 606 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |