Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021120.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021120.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021120.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021120.hg.1 |
TC10001621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021118.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021118.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021118.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021118.hg.1 |
TC10001621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179964.hg.1 |
TC1000011679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179966.hg.1 |
TC1000011679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000369937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000465409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000477994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
ENST00000486762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011679.hg.1 |
NM_024040.2 |
Refseq |