Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016178.hg.1 |
TC08001248.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016178.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016178.hg.1 |
ENST00000456290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016178.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171774.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171774.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171774.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171774.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171774.hg.1 |
TC0800010558.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
ENST00000361421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010558.hg.1 |
NM_014729.2 |
Refseq |