Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001132.1 | NP_001001132.1 | 14 - 15 | 1863 | 123 | |
NM_003024.2 | NP_003015.2 | 14 - 15 | 1863 | 123 | ||
XM_005261026.2 | XP_005261083.1 | 14 - 15 | 1941 | 123 | ||
XM_005261027.1 | XP_005261084.1 | 14 - 15 | 1941 | 123 | ||
XM_005261031.1 | XP_005261088.1 | 14 - 15 | 1941 | 123 | ||
XM_011529691.1 | XP_011527993.1 | 14 - 15 | 1830 | 123 | ||
XM_011529692.1 | XP_011527994.1 | 14 - 15 | 1830 | 123 | ||
XM_011529693.2 | XP_011527995.1 | 14 - 15 | 1941 | 123 | ||
XM_017028428.1 | XP_016883917.1 | 15 - 16 | 1861 | 123 | ||
XM_017028429.1 | XP_016883918.1 | 14 - 15 | 1830 | 123 | ||
XM_017028430.1 | XP_016883919.1 | 14 - 15 | 1830 | 123 | ||
XM_017028431.1 | XP_016883920.1 | 14 - 15 | 1941 | 123 | ||
XM_017028432.1 | XP_016883921.1 | 14 - 15 | 1941 | 123 | ||
XM_017028433.1 | XP_016883922.1 | 14 - 15 | 1941 | 123 | ||
XM_017028434.1 | XP_016883923.1 | 14 - 15 | 1941 | 123 | ||
XM_017028435.1 | XP_016883924.1 | 14 - 15 | 1830 | 123 | ||
XM_017028436.1 | XP_016883925.1 | 14 - 15 | 1830 | 123 | ||
XM_017028437.1 | XP_016883926.1 | 9 - 10 | 1281 | 123 | ||
XM_017028438.1 | XP_016883927.1 | 14 - 15 | 1830 | 123 | ||
XM_017028439.1 | XP_016883928.1 | 14 - 15 | 1941 | 123 | ||
XM_017028440.1 | XP_016883929.1 | 14 - 15 | 1830 | 123 | ||
XM_017028441.1 | XP_016883930.1 | 14 - 15 | 1941 | 123 | ||
GenBank mRNA | AF064243.1 | - | 14 - 15 | 1701 | 123 | |
AF064244.1 | - | 14 - 15 | 1701 | 123 | ||
AF114487.1 | - | 14 - 15 | 1863 | 123 | ||
AF114488.1 | - | 14 - 15 | 1863 | 123 | ||
AK027846.1 | - | 14 - 15 | 1830 | 123 | ||
AK074554.1 | - | 14 - 15 | 1799 | 123 | ||
AK300274.1 | - | 14 - 15 | 1838 | 123 | ||
BC013578.1 | - | 14 - 15 | 1726 | 123 | ||
BC020269.1 | - | 14 - 15 | 1776 | 123 | ||
BC058925.1 | - | 14 - 15 | 1890 | 123 | ||
BC073744.1 | - | 14 - 15 | 1805 | 123 | ||
BC116185.1 | - | 14 - 15 | 1674 | 40073780 | 123 | |
BC116186.2 | - | 14 - 15 | 1674 | 40073781 | 123 | |
BC117560.1 | - | 14 - 15 | 1843 | 7469144 | 123 | |
BX538175.1 | - | 9 - 10 | 1162 | 123 | ||
DQ340865.1 | - | 13 - 14 | 1591 | 123 | ||
DQ386455.1 | - | 13 - 14 | 1614 | 123 | ||
DQ679754.1 | - | 14 - 15 | 1680 | 123 | ||
DQ679755.1 | - | 14 - 15 | 1680 | 123 | ||
DQ679756.1 | - | 15 - 16 | 1569 | 123 | ||
DQ679757.1 | - | 14 - 15 | 1680 | 123 | ||
EU107386.1 | - | 14 - 15 | 1569 | 123 | ||
EU117382.1 | - | 13 - 14 | 1596 | 123 | ||
EU120733.1 | - | 13 - 14 | 1596 | 123 | ||
EU120734.1 | - | 13 - 14 | 1596 | 123 | ||
EU120735.1 | - | 14 - 15 | 1569 | 123 | ||
EU120736.1 | - | 14 - 15 | 1569 | 123 | ||
EU140799.1 | - | 14 - 15 | 1680 | 123 | ||
EU140800.1 | - | 13 - 14 | 1485 | 123 | ||
EU152331.1 | - | 13 - 14 | 1596 | 123 | ||
EU191901.1 | - | 13 - 14 | 1519 | 123 | ||
GU939616.1 | - | 9 - 10 | 1285 | 123 | ||
GU939617.1 | - | 9 - 10 | 1307 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |