Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |