Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003720.hg.1 |
TC01000232.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003720.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003720.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149601.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149601.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149601.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149601.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149601.hg.1 |
TC0100007105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375448.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000375453.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000467001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000468945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
ENST00000487048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007105.hg.1 |
NM_012387.2 |
Refseq |