Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006022.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006022.hg.1 |
ENST00000542955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006022.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006022.hg.1 |
TC17000499.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156730.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156730.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156730.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156730.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156730.hg.1 |
TC1700007796.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
ENST00000269593.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007796.hg.1 |
NM_001552.2 |
Refseq |