Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005275.hg.1 |
TC04000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005275.hg.1 |
ENST00000339336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005275.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005275.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007783.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
ENST00000339336.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
ENST00000504451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
NM_001286731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
NM_212557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153153.hg.1 |
TC0400007783.hg.1 |
Clariom D TC |