Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002332.2 | NP_002323.2 | 81 - 82 | 13047 | 63 | |
XM_017019303.1 | XP_016874792.1 | 82 - 83 | 13196 | 63 | ||
GenBank mRNA | AB209498.1 | - | 59 - 60 | 8792 | 63 | |
AK122840.1 | - | 9 - 10 | 3275 | 63 | ||
X13916.1 | - | 81 - 82 | 13047 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |