Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1211L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011511094.1 | XP_011509396.1 | 2 - 3 | 319 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |