Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010686.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010686.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010686.hg.1 |
TC17000828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000359939.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000549637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
ENST00000551294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008846.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
ENST00000389916.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
ENST00000547389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
NM_153209.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166042.hg.1 |
TC1700008846.hg.1 |
Clariom D TC |