Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
ENST00000330470.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
ENST00000332512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
ENST00000352483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
ENST00000542057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
ENST00000544709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005726.hg.1 |
TC21000476.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008222.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
NM_020639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
TC2100008222.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
ENST00000332512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
ENST00000352483.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155664.hg.1 |
NM_020639.2 |
Refseq |