Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000251607.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000280591.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000339437.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000397779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000397779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000397779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000397779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000397779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000402675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000413000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000420393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000434583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000465998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000465998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000465998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000465998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000469632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000469632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000469632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000469632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000469632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000482311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
ENST00000607052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_001302946.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006462.hg.1 |
NM_182916.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010395.hg.1 |
ENST00000450164.2 |
Ensembl |