Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099272.1 | NP_001092742.1 | 11 | 5927 | 135 | |
NM_001172418.1 | NP_001165889.1 | 11 | 5799 | 135 | ||
NM_052893.1 | NP_443125.1 | 12 | 6127 | 135 | ||
NM_152733.2 | NP_689946.2 | 10 | 5664 | 135 | ||
XM_011514279.2 | XP_011512581.1 | 13 | 6524 | 135 | ||
GenBank mRNA | BX537534.1 | - | 11 | 5623 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |