Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018557.2 | NP_061027.2 | 40 - 41 | 7399 | 97 | |
XM_017004341.1 | XP_016859830.1 | 40 - 41 | 11975 | 97 | ||
XM_017004342.1 | XP_016859831.1 | 11 - 12 | 1398 | 97 | ||
GenBank mRNA | AF176832.1 | - | 40 - 41 | 7399 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |