Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020766.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020766.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020766.hg.1 |
TC11001790.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020768.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020768.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020768.hg.1 |
TC11001790.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100182272.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182272.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182272.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182272.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182272.hg.1 |
TC1100010889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010889.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182270.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182270.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182270.hg.1 |
ENST00000287143.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182270.hg.1 |
NM_006093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182270.hg.1 |
TC1100010889.hg.1 |
Clariom D TC |