Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015178.hg.1 |
ENST00000251089.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015178.hg.1 |
ENST00000557179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015178.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015178.hg.1 |
TC14001334.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182895.hg.1 |
TC1400010771.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000554941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000556298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
ENST00000557497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010771.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182899.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182899.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182899.hg.1 |
ENST00000555079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182899.hg.1 |
NM_015305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182899.hg.1 |
TC1400010771.hg.1 |
Clariom D TC |