Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256959.1 | NP_001243888.1 | 7 - 8 | 753 | 109 | |
NM_001286.3 | NP_001277.1 | 8 - 9 | 819 | 109 | ||
GenBank mRNA | AK289999.1 | - | 8 - 9 | 724 | 109 | |
AK294764.1 | - | 7 - 8 | 608 | 109 | ||
BC117420.1 | - | 8 - 9 | 676 | 40125971 | 109 | |
BC117424.1 | - | 8 - 9 | 676 | 40125975 | 109 | |
D28475.2 | - | 8 - 9 | 704 | 109 | ||
X83378.1 | - | 8 - 9 | 678 | 109 | ||
X99473.1 | - | 8 - 9 | 652 | 109 | ||
X99474.1 | - | 8 - 9 | 652 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Homo sapiens Probe spans exons |