Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014718.hg.1 |
TC01000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014718.hg.1 |
ENST00000264126.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014718.hg.1 |
ENST00000406462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014718.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166519.hg.1 |
TC0100009333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000264126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000406462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000435987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000441735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
ENST00000446797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009333.hg.1 |
NM_013296.4 |
Refseq |