Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009813.hg.1 |
ENST00000615151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004440.hg.1 |
ENST00000283550.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004440.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004440.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004440.hg.1 |
TC13000409.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008072.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000283550.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000375699.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000414180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
ENST00000443541.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
NM_145248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154298.hg.1 |
TC1300008072.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009813.hg.1 |
ENST00000615151.1 |
Ensembl |