Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000361820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000479772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000479772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
ENST00000479772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011038.hg.1 |
NM_133445.2 |
Refseq |