Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004186.hg.1 |
TC09000391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004186.hg.1 |
ENST00000361671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004186.hg.1 |
ENST00000376040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004186.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152539.hg.1 |
TC0900007775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000361671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000376040.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
ENST00000416045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007775.hg.1 |
NM_024635.3 |
Refseq |