Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
TC06000592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000244670.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000326974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000332245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000394096.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000426116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
ENST00000432243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006792.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156090.hg.1 |
TC0600008066.hg.1 |
Clariom D TC |