Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003957.hg.1 |
ENST00000373433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003957.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003957.hg.1 |
TC20000278.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
NM_021215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
TC2000007300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
ENST00000373433.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
ENST00000462548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150468.hg.1 |
NM_021215.3 |
Refseq |