Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022193.hg.1 |
TC04001591.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022193.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022193.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011975.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182152.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182152.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182152.hg.1 |
ENST00000262999.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182152.hg.1 |
NM_021833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182152.hg.1 |
TC0400011975.hg.1 |
Clariom D TC |