Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031431.hg.1 |
TC03001725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031431.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031431.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191533.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191533.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191533.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191533.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191533.hg.1 |
TC0300012255.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
ENST00000296220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012255.hg.1 |
NM_022776.4 |
Refseq |