Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010905.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010905.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010905.hg.1 |
TC16000749.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168336.hg.1 |
TC1600009035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168337.hg.1 |
TC1600009035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000007390.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
ENST00000566296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009035.hg.1 |
NM_001001410.2 |
Refseq |