Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039654.hg.1 |
TC02002504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039654.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039654.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208241.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208241.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208241.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208241.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208241.hg.1 |
TC0200014834.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000355999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000461000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
ENST00000487143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014834.hg.1 |
NM_013233.2 |
Refseq |