Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052947.3 | NP_443179.3 | 8 - 9 | 5874 | 100 | |
XM_011525801.2 | XP_011524103.1 | 8 - 9 | 6107 | 100 | ||
XM_011525802.2 | XP_011524104.1 | 6 - 7 | 6494 | 100 | ||
XM_017025535.1 | XP_016881024.1 | 8 - 9 | 6108 | 100 | ||
XM_017025536.1 | XP_016881025.1 | 8 - 9 | 6108 | 100 | ||
XM_017025537.1 | XP_016881026.1 | 8 - 9 | 6109 | 100 | ||
XM_017025538.1 | XP_016881027.1 | 5 - 6 | 4992 | 100 | ||
XM_017025539.1 | XP_016881028.1 | 5 - 6 | 3740 | 100 | ||
GenBank mRNA | AL832018.2 | - | 5 - 6 | 4206 | 100 | |
AY044450.1 | - | 5 - 6 | 3941 | 100 | ||
BX647446.1 | - | 4 - 5 | 3352 | 100 | ||
BX647639.1 | - | 5 - 6 | 4740 | 100 | ||
BX647796.1 | - | 8 - 9 | 5861 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons |