Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009000.hg.1 |
ENST00000477363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009000.hg.1 |
ENST00000477363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011107.hg.1 |
ENST00000497334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007489.hg.1 |
ENST00000584631.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007031.hg.1 |
ENST00000439099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007595.hg.1 |
ENST00000413846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007540.hg.1 |
ENST00000437667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000329092.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000329092.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000329092.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000329092.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000429437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000429437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000429437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000429437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000439255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000442341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000442341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000442341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000442341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000464167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000474640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000485439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000485439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000485439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000485439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000496582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
ENST00000496582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
NM_000996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
NM_000996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
NM_000996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
NM_000996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010076.hg.1 |
NM_000996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007572.hg.1 |
ENST00000430869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009335.hg.1 |
ENST00000489757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011794.hg.1 |
ENST00000468042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009262.hg.1 |
ENST00000586202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009262.hg.1 |
ENST00000586202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010687.hg.1 |
ENST00000482557.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009110.hg.1 |
ENST00000484185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008629.hg.1 |
ENST00000404414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009307.hg.1 |
ENST00000425552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008173.hg.1 |
ENST00000478069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007632.hg.1 |
ENST00000469738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013293.hg.1 |
ENST00000414680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009120.hg.1 |
ENST00000444401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007159.hg.1 |
ENST00000424997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016243.hg.1 |
ENST00000440329.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006594.hg.1 |
ENST00000427308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011612.hg.1 |
ENST00000402490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011612.hg.1 |
ENST00000402490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007280.hg.1 |
ENST00000462203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006964.hg.1 |
ENST00000468671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011541.hg.1 |
ENST00000412007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007585.hg.1 |
ENST00000459730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009826.hg.1 |
ENST00000469688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017007.hg.1 |
ENST00000427282.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007467.hg.1 |
ENST00000418340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018077.hg.1 |
ENST00000413092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012736.hg.1 |
ENST00000476981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011643.hg.1 |
ENST00000469425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009001.hg.1 |
ENST00000495857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016243.hg.1 |
ENST00000440329.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017007.hg.1 |
ENST00000427282.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008173.hg.1 |
ENST00000478069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009001.hg.1 |
ENST00000495857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011541.hg.1 |
ENST00000412007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011643.hg.1 |
ENST00000469425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009120.hg.1 |
ENST00000444401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009110.hg.1 |
ENST00000484185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300173374.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300173374.hg.1 |
ENST00000448864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300173374.hg.1 |
TC0300010076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018077.hg.1 |
ENST00000413092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007159.hg.1 |
ENST00000424997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008629.hg.1 |
ENST00000404414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011612.hg.1 |
ENST00000402490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011612.hg.1 |
ENST00000402490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009335.hg.1 |
ENST00000489757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011107.hg.1 |
ENST00000497334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007540.hg.1 |
ENST00000437667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007585.hg.1 |
ENST00000459730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006964.hg.1 |
ENST00000468671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007595.hg.1 |
ENST00000413846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012736.hg.1 |
ENST00000476981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007280.hg.1 |
ENST00000462203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009826.hg.1 |
ENST00000469688.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007572.hg.1 |
ENST00000430869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009000.hg.1 |
ENST00000477363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009000.hg.1 |
ENST00000477363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011794.hg.1 |
ENST00000468042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007031.hg.1 |
ENST00000439099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007489.hg.1 |
ENST00000584631.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007632.hg.1 |
ENST00000469738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010687.hg.1 |
ENST00000482557.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009262.hg.1 |
ENST00000586202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009262.hg.1 |
ENST00000586202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009307.hg.1 |
ENST00000425552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013293.hg.1 |
ENST00000414680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006594.hg.1 |
ENST00000427308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007467.hg.1 |
ENST00000418340.1 |
Ensembl |