Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042605.hg.1 |
TC02002649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042605.hg.1 |
ENST00000295049.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042605.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212492.hg.1 |
TC0200015350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000295049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000429081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000454447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
ENST00000494346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015350.hg.1 |
NM_144629.2 |
Refseq |